Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111179.4 | NP_186959.2 | 6 - 7 | 726 | 127 | |
GenBank mRNA | AY140054.1 | - | 6 - 7 | 615 | 127 | |
BT010375.1 | - | 6 - 7 | 477 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |