Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118225.5 | NP_193839.4 | 12 - 13 | 2587 | 103 | |
GenBank mRNA | AF515728.1 | - | 12 - 13 | 2454 | 103 | |
AY081328.1 | - | 12 - 13 | 2500 | 103 | ||
BT010547.1 | - | 12 - 13 | 2404 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |