Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118287.5 | NP_193898.3 | 10 - 11 | 2738 | 97 | |
GenBank mRNA | AY557186.1 | - | 10 - 11 | 2216 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |