Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203866.1 | NP_001190795.1 | 3 - 4 | 334 | 82 | |
NM_118320.3 | NP_193930.1 | 2 - 3 | 341 | 82 | ||
GenBank mRNA | AF428445.1 | - | 2 - 3 | 293 | 82 | |
AY054175.1 | - | 2 - 3 | 294 | 82 | ||
AY062665.1 | - | 2 - 3 | 285 | 82 | ||
AY093319.1 | - | 2 - 3 | 209 | 82 | ||
AY103313.1 | - | 2 - 3 | 209 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |