Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102760.3 | NP_564351.1 | 2 - 3 | 1455 | 100 | |
NM_179399.3 | NP_849730.1 | 3 - 4 | 1328 | 100 | ||
GenBank mRNA | AY035008.1 | - | 2 - 3 | 1427 | 100 | |
AY057672.1 | - | 3 - 4 | 1308 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |