Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102775.4 | NP_174327.1 | 2 - 3 | 306 | 93 | |
GenBank mRNA | AY039848.1 | - | 2 - 3 | 225 | 93 | |
AY074826.1 | - | 2 - 3 | 164 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |