Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332962.1 | NP_001320669.1 | 10 - 11 | 1920 | 63 | |
NM_102881.4 | NP_174427.3 | 11 - 12 | 1505 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |