Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XLG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198196.1 | NP_001185125.1 | 8 - 9 | 2246 | 55 | |
NM_001332988.1 | NP_001319126.1 | 8 - 9 | 2358 | 55 | ||
NM_001332989.1 | NP_001320661.1 | 8 - 9 | 2448 | 55 | ||
NM_001332990.1 | NP_001320662.1 | 7 - 8 | 2482 | 55 | ||
NM_102929.2 | NP_174475.1 | 9 - 10 | 2268 | 55 | ||
NM_179406.2 | NP_849737.1 | 8 - 9 | 2321 | 55 | ||
GenBank mRNA | AK229561.1 | - | 8 - 9 | 2241 | 55 | |
AK317243.1 | - | 8 - 9 | 2092 | 55 | ||
AK317663.1 | - | 7 - 8 | 2011 | 55 | ||
GQ229390.1 | - | 9 - 10 | 2271 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |