Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102967.3 | NP_174512.1 | 6 - 7 | 2024 | 64 | |
GenBank mRNA | AY099548.1 | - | 6 - 7 | 1963 | 64 | |
BT002115.1 | - | 6 - 7 | 1941 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |