Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120061.2 | NP_568050.1 | 4 - 5 | 1321 | 94 | |
GenBank mRNA | AY059825.1 | - | 4 - 5 | 1238 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |