Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337010.1 | NP_001324372.1 | 2 - 3 | 608 | 108 | |
NM_180051.3 | NP_850382.1 | 3 - 4 | 908 | 108 | ||
GenBank mRNA | AF474176.1 | - | 3 - 4 | 725 | 108 | |
BT046199.1 | - | 2 - 3 | 425 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |