Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342587.1 | NP_001330013.1 | 12 - 13 | 2855 | 99 | |
NM_001342588.1 | NP_001330014.1 | 13 - 14 | 2812 | 99 | ||
NM_120218.2 | NP_195760.1 | 13 - 14 | 2576 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |