Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342651.1 | NP_001318457.1 | 9 - 10 | 4580 | 90 | |
NM_001342652.1 | NP_001331954.1 | 9 - 10 | 4603 | 90 | ||
NM_120309.2 | NP_195851.2 | 8 - 9 | 4245 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |