Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120449.3 | NP_195988.1 | 1 - 2 | 706 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |