Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120473.3 | NP_196011.1 | 6 - 7 | 1419 | 112 | |
GenBank mRNA | AY059727.1 | - | 6 - 7 | 1414 | 112 | |
AY091365.1 | - | 6 - 7 | 1352 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |