Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127402.4 | NP_179436.3 | 3 - 4 | 377 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |