Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127427.3 | NP_179461.1 | 2 - 3 | 392 | 88 | |
GenBank mRNA | AF424550.1 | - | 2 - 3 | 377 | 88 | |
AY058103.1 | - | 2 - 3 | 377 | 88 | ||
AY065093.1 | - | 2 - 3 | 384 | 88 | ||
BT002632.1 | - | 2 - 3 | 359 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |