Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035689.2 | NP_001030766.1 | 14 - 15 | 1447 | 143 | |
NM_001035690.1 | NP_001030767.1 | 15 - 16 | 1506 | 143 | ||
NM_001338758.1 | NP_001327446.1 | 14 - 15 | 1611 | 143 | ||
NM_001338759.1 | NP_001327445.1 | 13 - 14 | 1056 | 143 | ||
NM_113456.4 | NP_189186.1 | 15 - 16 | 1510 | 143 | ||
NM_180413.2 | NP_850744.1 | 15 - 16 | 1309 | 143 | ||
GenBank mRNA | AF360151.1 | - | 15 - 16 | 1374 | 143 | |
AY054269.1 | - | 15 - 16 | 1358 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |