Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343401.1 | NP_001318568.1 | 2 - 3 | 1123 | 118 | |
NM_001343402.1 | NP_001331477.1 | 2 - 3 | 499 | 118 | ||
NM_121550.3 | NP_568315.1 | 3 - 4 | 457 | 118 | ||
NM_180493.4 | NP_850824.1 | 3 - 4 | 787 | 118 | ||
GenBank mRNA | AK228599.1 | - | 3 - 4 | 457 | 118 | |
AY086424.1 | - | 3 - 4 | 451 | 118 | ||
BT005773.1 | - | 3 - 4 | 457 | 118 | ||
BT006108.1 | - | 2 - 3 | 302 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |