Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180596.2 | NP_850927.1 | 12 - 13 | 1636 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |