Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342847.1 | NP_001332731.1 | 4 - 5 | 831 | 84 | |
NM_120676.3 | NP_196213.2 | 4 - 5 | 831 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |