Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120717.5 | NP_196252.1 | 5 - 6 | 712 | 84 | |
GenBank mRNA | AF372941.1 | - | 5 - 6 | 645 | 84 | |
AY113163.1 | - | 5 - 6 | 584 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |