Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBOHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203321.1 | NP_001190250.1 | 6 - 7 | 1328 | 93 | |
NM_120821.2 | NP_196356.1 | 5 - 6 | 1469 | 93 | ||
GenBank mRNA | BT003857.1 | - | 5 - 6 | 1469 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |