Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036226.2 | NP_001031303.1 | 3 - 4 | 579 | 80 | |
NM_001198511.1 | NP_001185440.1 | 3 - 4 | 336 | 80 | ||
NM_106631.5 | NP_178100.3 | 3 - 4 | 528 | 80 | ||
NM_180652.3 | NP_850983.1 | 3 - 4 | 322 | 80 | ||
GenBank mRNA | AK317667.1 | - | 2 - 3 | 249 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |