Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATTPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344937.1 | NP_001332431.1 | 9 - 10 | 1353 | 105 | |
NM_001344938.1 | NP_001332432.1 | 9 - 10 | 1246 | 105 | ||
NM_124525.2 | NP_199959.2 | 8 - 9 | 1117 | 105 | ||
NM_180840.2 | NP_851171.1 | 9 - 10 | 1357 | 105 | ||
NM_203193.3 | NP_974922.1 | 8 - 9 | 1670 | 105 | ||
GenBank mRNA | AF007778.1 | - | 9 - 10 | 1244 | 105 | |
AY093147.1 | - | 8 - 9 | 1117 | 105 | ||
BT008467.1 | - | 8 - 9 | 938 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |