Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124600.3 | NP_200034.1 | 3 - 4 | 464 | 113 | |
NM_180844.2 | NP_851175.1 | 3 - 4 | 346 | 113 | ||
GenBank mRNA | AK227533.1 | - | 3 - 4 | 328 | 113 | |
BT004590.1 | - | 2 - 3 | 229 | 113 | ||
U46924.1 | - | 3 - 4 | 435 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |