Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125782.3 | NP_568979.1 | 2 - 3 | 513 | 125 | |
NM_180927.4 | NP_851258.2 | 2 - 3 | 566 | 125 | ||
NM_180928.2 | NP_851259.1 | 2 - 3 | 555 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |