Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125820.3 | NP_201229.1 | 1 - 2 | 618 | 101 | |
NM_180931.2 | NP_851262.1 | 1 - 2 | 618 | 101 | ||
GenBank mRNA | AY086951.1 | - | 1 - 2 | 438 | 101 | |
AY322527.1 | - | 1 - 2 | 402 | 101 | ||
BT002395.1 | - | 1 - 2 | 436 | 101 | ||
BT010372.1 | - | 1 - 2 | 402 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |