Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339535.1 | NP_001326349.1 | 2 - 3 | 1080 | 94 | |
NM_001339536.1 | NP_001326348.1 | 2 - 3 | 1011 | 94 | ||
NM_115044.5 | NP_190753.2 | 2 - 3 | 1080 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |