Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115145.2 | NP_190853.1 | 11 - 12 | 2019 | 79 | |
GenBank mRNA | U86700.1 | - | 11 - 12 | 1938 | 79 | |
Y07917.1 | - | 1948 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |