Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIP37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115277.5 | NP_190985.1 | 6 - 7 | 633 | 72 | |
GenBank mRNA | AF084570.1 | - | 6 - 7 | 515 | 72 | |
AY063940.1 | - | 6 - 7 | 540 | 72 | ||
AY113943.1 | - | 6 - 7 | 449 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |