Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATVPS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161207.1 | NP_001154679.1 | 5 - 6 | 559 | 122 | |
NM_115343.4 | NP_567009.1 | 5 - 6 | 672 | 122 | ||
GenBank mRNA | AF357527.1 | - | 5 - 6 | 492 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |