Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115496.2 | NP_567037.1 | 3 - 4 | 339 | 96 | |
GenBank mRNA | AF305722.1 | - | 3 - 4 | 294 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |