Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339858.1 | NP_001319779.1 | 14 - 15 | 1693 | 100 | |
NM_001339859.1 | NP_001325644.1 | 14 - 15 | 1693 | 100 | ||
NM_001339860.1 | NP_001325645.1 | 14 - 15 | 2458 | 100 | ||
NM_001339861.1 | NP_001325651.1 | 14 - 15 | 2512 | 100 | ||
NM_001339862.1 | NP_001325648.1 | 14 - 15 | 2127 | 100 | ||
NM_001339863.1 | NP_001325647.1 | 14 - 15 | 2165 | 100 | ||
NM_001339864.1 | NP_001325650.1 | 14 - 15 | 1673 | 100 | ||
NM_001339865.1 | NP_001325649.1 | 13 - 14 | 1829 | 100 | ||
NM_001339866.1 | NP_001325646.1 | 13 - 14 | 1867 | 100 | ||
NM_115601.3 | NP_567048.1 | 14 - 15 | 1731 | 100 | ||
GenBank mRNA | AK318626.1 | - | 13 - 14 | 1684 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |