Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124279.5 | NP_199713.2 | 7 - 8 | 937 | 70 | |
NM_203184.2 | NP_974913.1 | 7 - 8 | 871 | 70 | ||
GenBank mRNA | BT030319.1 | - | 7 - 8 | 787 | 70 | |
BT030323.1 | - | 7 - 8 | 787 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |