Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125859.4 | NP_568990.1 | 6 - 7 | 779 | 142 | |
NM_203262.3 | NP_974991.1 | 6 - 7 | 779 | 142 | ||
NM_203263.2 | NP_974992.1 | 6 - 7 | 694 | 142 | ||
GenBank mRNA | AF375435.1 | - | 6 - 7 | 715 | 142 | |
AY074827.1 | - | 6 - 7 | 646 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |