Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333051.1 | NP_001319134.1 | 6 - 7 | 639 | 151 | |
NM_001333052.1 | NP_001321942.1 | 5 - 6 | 718 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |