Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333320.1 | NP_001321802.1 | 6 - 7 | 844 | 100 | |
NM_103724.2 | NP_175261.1 | 6 - 7 | 844 | 100 | ||
GenBank mRNA | AK228479.1 | - | 6 - 7 | 697 | 100 | |
U95143.1 | - | 6 - 7 | 840 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |