Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203212.1 | NP_001190141.1 | 20 - 21 | 2802 | 85 | |
NM_001340039.1 | NP_001327362.1 | 20 - 21 | 2795 | 85 | ||
NM_001340040.1 | NP_001327363.1 | 19 - 20 | 2829 | 85 | ||
NM_115897.5 | NP_191592.5 | 19 - 20 | 2800 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |