Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340050.1 | NP_001319806.1 | 2 - 3 | 354 | 65 | |
NM_001340051.1 | NP_001325988.1 | 1 - 2 | 132 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |