Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121311.5 | NP_196812.1 | 1 - 2 | 442 | 77 | |
GenBank mRNA | AF452174.1 | - | 1 - 2 | 260 | 77 | |
BT002426.1 | - | 1 - 2 | 391 | 77 | ||
BT006530.1 | - | 1 - 2 | 260 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |