Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085110.1 | NP_001078579.1 | 6 - 7 | 493 | 107 | |
NM_121374.4 | NP_196875.1 | 7 - 8 | 777 | 107 | ||
GenBank mRNA | AF090372.1 | - | 7 - 8 | 444 | 107 | |
AF195648.1 | - | 7 - 8 | 455 | 107 | ||
AY120716.1 | - | 7 - 8 | 469 | 107 | ||
BT000058.1 | - | 6 - 7 | 402 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |