Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121377.5 | NP_196878.2 | 17 - 18 | 1635 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |