Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT3;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121407.3 | NP_196908.1 | 1 - 2 | 478 | 67 | |
GenBank mRNA | AY058848.1 | - | 1 - 2 | 429 | 67 | |
AY143869.1 | - | 1 - 2 | 324 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |