Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125758.2 | NP_001119230.1 | 5 - 6 | 996 | 107 | |
NM_121585.4 | NP_568322.1 | 5 - 6 | 907 | 107 | ||
GenBank mRNA | AK118608.1 | - | 5 - 6 | 824 | 107 | |
BT006224.1 | - | 5 - 6 | 510 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |