Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121605.3 | NP_197104.1 | 6 - 7 | 939 | 70 | |
GenBank mRNA | AY074263.1 | - | 6 - 7 | 728 | 70 | |
BT002366.1 | - | 6 - 7 | 609 | 70 | ||
FJ708777.1 | - | 6 - 7 | 609 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |