Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AILP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343590.1 | NP_001330082.1 | 3 - 4 | 459 | 106 | |
NM_121919.3 | NP_197415.1 | 3 - 4 | 696 | 106 | ||
GenBank mRNA | AY039946.1 | - | 3 - 4 | 692 | 106 | |
AY113899.1 | - | 3 - 4 | 567 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |