Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121927.3 | NP_197423.1 | 10 - 11 | 1170 | 58 | |
GenBank mRNA | AF370503.1 | - | 10 - 11 | 1169 | 58 | |
BT008884.1 | - | 10 - 11 | 1114 | 58 | ||
U72290.1 | - | 10 - 11 | 1127 | 58 | ||
X73367.2 | - | 1127 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |