Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202728.1 | NP_001189657.1 | 2 - 3 | 474 | 116 | |
NM_128803.4 | NP_180803.1 | 1 - 2 | 332 | 116 | ||
GenBank mRNA | BT029168.1 | - | 1 - 2 | 234 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |