Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202731.1 | NP_001189660.1 | 17 - 18 | 2197 | 96 | |
NM_128841.5 | NP_565755.1 | 17 - 18 | 2267 | 96 | ||
GenBank mRNA | AJ270305.1 | - | 17 - 18 | 2171 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |